Learn More
Promega Restriction Enzymes - Sal I Blue/White Cloning and Genome Qualified (D) DFS Item
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications. (G/TCGA C C AGCT\G)
Supplier:  Promega R6055
 View more versions of this product
Catalog No. PR-R6055
Only null left
Explore available promotions
Description
Description
  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
Specifications
Specifications
10 U/μL
37°C
Provides a higher level of quality control for Enzymes used in cloning applications
-30°C to -10°C
G.TCGAC CAGCT.G
Liquid
1 x 1mL 10X Buffer D, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 10000U Sall
7.4
10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
10,000 U
SalI
Product Suggestions
Product Suggestions