Learn More
Promega Replacement Products for the Wizard™ Magnetic DNA Purification System for Food
Components of the Wizard™ Magnetic DNA Purification System for Food, which is intended for isolating genomic DNA from a variety of food samples including: corn seeds, corn meal, soy beans, soy flour, soy milk, corn oil, soybean oil, canola oil, etc.
Supplier: Promega A8191
Description
- Lysis Buffers Used in Protocol for Isolating DNA from Food
- Components of the Wizard(R) Magnetic DNA Purification System for Food available as standalone buffers
Specifications
| 15°C to 30°C | |
| Purifies DNA from a variety of food samples including corn seeds, cornmeal, soybeans, Soy flour and Soy milk | |
| 100 mL |
| Wizard™ Magnetic DNA purification system for food | |
| Lysis buffer; A |