missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Replacement Products for the Wizard™ Magnetic DNA Purification System for Food
Components of the Wizard™ Magnetic DNA Purification System for Food, which is intended for isolating genomic DNA from a variety of food samples including: corn seeds, corn meal, soy beans, soy flour, soy milk, corn oil, soybean oil, canola oil, etc.
$197.64 - $795.83
Specifications
| Content And Storage | 15°C to 30°C |
|---|---|
| For Use With (Equipment) | Wizard™ Magnetic DNA purification system for food |
| For Use With (Application) | Purifies DNA from a variety of food samples including corn seeds, cornmeal, soybeans, Soy flour and Soy milk |
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Product Type | Price | Quantity | ||||
PRA8191
|
Promega
A8191 |
100 mL | Lysis buffer; A |
Each for $197.64
|
|
||||
PRZ3191
|
Promega
Z3191 |
100 mL | Lysis buffer; B |
Each for $433.49
|
|
||||
PRZ3201
|
Promega
Z3201 |
150 mL | Precipitation solution |
Each for $795.83
|
|
||||
Description
- Lysis Buffers Used in Protocol for Isolating DNA from Food
- Components of the Wizard(R) Magnetic DNA Purification System for Food available as standalone buffers
Specifications
| 15°C to 30°C | |
| Wizard™ Magnetic DNA purification system for food | |
| Purifies DNA from a variety of food samples including corn seeds, cornmeal, soybeans, Soy flour and Soy milk |