Learn More
Promega ReliaPrep™ gDNA Tissue Miniprep System DFS Item
The ReliaPrep gDNA Tissue Miniprep System provides a complete, ready-to-use method for purification of gDNA from up to 25mg of tissue, a buccal swab, or a 1cm mouse tail snip, obtaining intact gDNA without the use of ethanol washes or precipitations.
Supplier:  Promega A2052
 View more versions of this product
Catalog No. PRA2052
Only null left
Explore available promotions
Description
Description
  • Manual DNA Extraction from 25mg Tissue, 1 Buccal Swab, or 1cm Mouse Tail
  • No ethanol, eliminating alcohol inhibition and carryover.
  • Good recovery and purity in as little as 50μL elution
  • Consistent results from run-to-run and user-to-user
Specifications
Specifications
15°C to 30°C
Purifies genomic DNA from up to 25mg of tissue, a buccal swab or a 1cm Mouse tail snip, obtaining intact genomic DNA without the use of Ethanol washes or precipitations
2 x 68.75mL Binding Buffer, 1 x 412.5mL Column wash solution, 5 x 50 ReliaPrep™ binding columns, 1 x 5.5mL Proteinase K solution, 1 x 100mL Cell Lysis Buffer, 5 x 200 collection tubes, 1 x 25mL tail Lysis Buffer, 6 x 1mL RNase A solution
250 Preps
Product Suggestions
Product Suggestions