missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega ReliaPrep™ gDNA Tissue Miniprep System
Description
- Manual DNA Extraction from 25mg Tissue, 1 Buccal Swab, or 1cm Mouse Tail
- No ethanol, eliminating alcohol inhibition and carryover.
- Good recovery and purity in as little as 50μL elution
- Consistent results from run-to-run and user-to-user
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| For Use With (Application) | Purifies genomic DNA from up to 25mg of tissue, a buccal swab or a 1cm Mouse tail snip, obtaining intact genomic DNA without the use of Ethanol washes or precipitations |
| Includes | 1 x 50mL Binding Buffer, 1 x 165mL Column wash solution, 2 x 50 ReliaPrep™ binding columns, 2 x 1.1mL Proteinase K solution, 1 x 40mL Cell Lysis Buffer, 10 x 40 collection tubes, 1 x 10mL tail Lysis Buffer, 1 x 250μL RN |
| No. of Reactions | 100 Preps |