Learn More
Promega ReliaPrep™ FFPE gDNA Miniprep System
The ReliaPrep FFPE gDNA Miniprep System provides a complete, all-inclusive method for purifying quality genomic DNA from formalin-fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion.
Supplier: Promega A2352
Description
- Intact, Amplifiable DNA from FFPE Tissue
- Minimal preparation time: simply add ethanol and go!
- Safely deparaffinize without using harsh organic solvents
- No overnight digestion required
Identifiants chimiques
| 15 to 30°C | |
| Purifying genomic dna from formalin-fixed paraffin-embedded tissue | |
| Ffpe Gdna Miniprep System |
Spécifications
| 15 to 30°C | |
| 5mL mineral oil, 20mL lysis buffer, 2 x 1.1mL proteinase K, 3.25mL BL buffer, 30mL wash solution, 1mL RNase A, 15mL elution buffer, collection tubes (50 tubes/package), ReliaPrep™ FFPE binding columns (50 columns/package) | |
| 100 Reactions |
| Purifying genomic dna from formalin-fixed paraffin-embedded tissue | |
| Ffpe Gdna Miniprep System |