Learn More
Promega ReadyAmp™ Genomic DNA Purification System
System for purification of single-stranded DNA from whole blood or blood stains. DNA may be used directly in amplification reactions without further manipulation. No organic extraction or ethanol precipitation.
Supplier:  Promega A7710
Encompass
Numéro de catalogue. PRA7710
Il en reste null
Description
Description
  • Single-Stranded DNA from Whole Blood or Blood Stains
  • Removes PCR inhibitors
  • Isolated DNA can be used directly in PCR
Spécifications
Spécifications
15°C to 30°C
20mL ReadyAmp™ genomic DNA purification Resin, 100mL Nuclease-free Water, autoclaved Magnetic stir bar, protocol
System
Produces single-stranded DNA (ssDNA) from whole Blood or Blood stains
100
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS