Learn More
Promega ProTEV Plus Protease
ProTEV Plus is an improved 48kDa version of the Nla protease from tobacco etch virus (TEV) that has been engineered to be more stable than native TEV protease for prolonged enzymatic activity.
Supplier: Promega V6102
Description
- Engineered Protease with Greater Stability, Prolonged Activity Over Native TEV Protease
- Active over broad pH (5.5-8.5) and temperature (4-30°C) range
- Cleaves protein fusions with the ENLYFQ(G/S) sequence
- Use with in-solution and on-column digestions
- Supplied at a concentration of 5u/μL
Specifications
| 5 U/μL | |
| HQ tag located at N-terminus to be immobilized on affinity resins | |
| 8000 U |
| Cleavage of affinity tags from fusion Proteins after Protein purification, removal of HaloTag™ Protein during Protein purification/expression of used Proteins of interest | |
| -30°C to -10°C | |
| Protease |