missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Proteasome-GLO™ Cell-Based Assays
Description
Comprend
Buffer, substrate and luciferin detection reagent
- Homogeneous, Luminescent Assays that Measure Protease Activities in Cultured Cells
- Simple add-mix-measure protocol
- Chymotrypsin-Like, Trypsin-Like and Caspase-Like activity data directly from cellular environment
- Results within 10-30 minutes after reagent addition
Order Info
Assay system also available separately (Cat. No. PRG1200).
Spécifications
Spécifications
| Buffer | Proteasome-Glo™ cell-based buffer |
| Product Type | Caspase-like cell-based assay kit |
| Quantity | 5 x 10 mL |
| Includes | Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent |
| Content And Storage | −30 to −10°C |
| For Use With (Application) | Measures Caspase-like Protease activity in cultured cells |