missing translation for 'onlineSavingsMsg'
Learn More

Promega PCR Markers

Numéro de catalogue. prg3161
Encompass
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
PRG3161 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PRG3161 Fournisseur Promega Code fournisseur G3161
Il en reste null

PCR Markers: 6 bands of 50, 150, 300, 500, 750 and 1,000bp.

  • Six Equally Intense Bands of 50, 150, 300, 500, 750 and 1,000bp
  • Load 5μL/lane
  • Supplied at a concentration of ∼0.06μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Spécifications

Color Blue, Orange
Concentration 0.06 μg/μL
Form Liquid
Formulation 1 x 250μL PCR marker, 1 x 1mL blue/orange 6X loading dye
For Use With (Application) Sizing PCR products and restriction fragments
No. of Bands 6
Quantity 250 μL
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker