missing translation for 'onlineSavingsMsg'
Learn More

Promega Magne™ Protein G Magnetic Affinity Beads

Catalog No. PRG7473
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG7473 50 mL
PRG7471 1 mL
PRG7472 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG7473 Supplier Promega Supplier No. G7473
Only null left

Magne Protein G & Magne Protein A Beads are magnetic affinity beads with high specificity and capacity for purification of immunoglobulins from cell culture media, ascites and serum. Iron encapsulated in macroporous cellulose with low nonspecific binding.

  • High Specificity, High Capacity Magnetic Affinity Beads for Antibody Capture
  • Binding capacities of >25mg/mL depending on antibody species, isotype
  • Minimize purification losses and increase sample throughput with exceptional magnetic properties

Specifications

Binding Property ≤25mg/mL
Formulation 20% Slurry in 20% Ethanol
Content And Storage 2°C to 10°C
Particle Size 30 to 80 μm
Quantity 50 mL
For Use With (Application) Antibody purification from Serum, ascites and cell-media samples, auto mated, high-throughput Antibody purification, Antibody purification and concentration from small-volume or dilute source material
Type Magnetic Affinity Bead