Learn More
Promega MagaZorb™ DNA Miniprep Kits DFS Item
The MagaZorb DNA Kit provides an easy, fast and cost-effective technique for isolating PCR-quality DNA.
Supplier:  Promega PRMB1008
 View more versions of this product
Catalog No. PRMB1008
Only null left
Explore available promotions
Description
Description
  • Fast Isolation of PCR-quality DNA from Various Samples
  • All reagents provided, no reagent preparation required
  • Magnetic purification method increases throughput and reproducibility
  • No organic solvents
Specifications
Specifications
15°C to 30°C
16mL Proteinase K solution, 160mL Lysis Buffer, 400mL Binding Buffer, 16mL MagaZorb™ reagent, 4 x 400mL Wash Buffer, 480mL Elution Buffer
Kit
Isolating PCR-quality DNA
800 Preps
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
U.S. Patents 6,855,499 and other patents pending.