Learn More
Promega Luciferase Assay System Components DFS Item
For use with Promega Luciferase Assay Systems (see cat. no. PRE1500)
Supplier:  Promega E1483
 View more versions of this product
Catalog No. PR-E1483
Only null left
Explore available promotions
Description
Description
  • Ready-to-Use Reagent for Detection of Firefly Luciferase Activity
  • Convenient premixed reagent containing Luciferase Assay Substrate and Buffer, used in the Luciferase Assay System (Cat.# E1500, E1501)
  • Linear over eight or more orders of magnitude of enzyme concentration
  • Sensitive to 10-20 moles of luciferase
  1. A premix of Luciferase Assay Substrate and Luciferase Assay Buffer.
  2. U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
  3. The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  4. Certain applications of this product may require licenses from others.
Specifications
Specifications
Luciferase Assay Reagent
<-65°C
Liquid
8
100 mL
Product Suggestions
Product Suggestions