missing translation for 'onlineSavingsMsg'
Learn More

Promega Luciferase Assay System Components

Catalog No. p-4907533
Encompass
Change view
Click to view available options
Quantity:
30 mL
100 mL
Product Type:
Luciferase Assay Reagent
Luciferase Cell Culture Lysis 5X Reagent
Reporter Lysis 5X Buffer
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRE1531 30 mL Luciferase Cell Culture Lysis 5X Reagent
PRE3971 30 mL Reporter Lysis 5X Buffer
PRE1483 100 mL Luciferase Assay Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRE1531 Supplier Promega Supplier No. E1531
Only null left

For use with Promega Luciferase Assay Systems (see cat. no. PRE1500)

  • Rapid Lysis of Many Cell Types
  • Used with the Luciferase Assay System (Cat.# E1500) for reporter quantitation in mammalian cells
  • Provides rapid lysis of adherent and nonadherent mammalian cells, bacterial cells, plant cells and tissue homogenates

  1. A premix of Luciferase Assay Substrate and Luciferase Assay Buffer.
  2. U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents and patents pending.
  3. The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
  4. Certain applications of this product may require licenses from others.

Specifications

Product Type Luciferase Cell Culture Lysis 5X Reagent
Concentration 5 X
Includes 1 x 30 mL 5X Cell culture Lysis Reagent
pH 7.8
Content And Storage -30°C to -10°C
Quantity 30 mL
Form Liquid