missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Lambda DNA/HindIII Conventional DNA Digest Marker
Description
Comprend
6X Blue/Orange Loading Dye
- Eight DNA Fragments of 125-23,130bp
- Load 1μL/lane
- Supplied at a concentration of 0.5μg/μL
- Supplied with 6X Blue/Orange Loading Dye
Spécifications
Spécifications
| Buffer | 10mM Tris HCl pH 7.8 |
| Color | Blue, Orange |
| Concentration | 0.5 μg/μL |
| Form | Liquid |
| Formulation | 100μg lambda DNA/HindIII marker, 1mL blue/orange 6X loading dye |
| For Use With (Application) | Separation, size determination and quantitation of DNA and RNA fragments |
| No. of Bands | 8 |
| Quantity | 100 μg |
| Storage Requirements | −30 to −10°C |
| Product Type | DNA Molecular Weight Marker |