missing translation for 'onlineSavingsMsg'
Learn More

Promega Kinase-Glo™ Luminescent Kinase Assay

Catalog No. PRV6713
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
10 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6713 100 mL
PRV6711 10 mL
PRV6712 10 x 10 mL
PRV6714 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRV6713 Supplier Promega Supplier No. V6713

The Kinase-Glo™ Luminescent Kinase Assays are homogeneous non-radioactive systems for determining the activity of purified kinases by quantifying the amount of ATP remaining in solution following a kinase reaction.

  • Monitors Activity of Purified Kinases by Quantifying ATP
  • Luminescent signal is inversely proportional to the amount of kinase activity
  • Three kit formats: Linear to 10microM (Kinase-Glo(R)); 100microM (Kinase-Glo(R) Plus) or 500microM (Kinase-Glo(R) Max) ATP

  1. U.S. Patent 6,602,677, Australian Patent 754312 and other patents pending.
  2. The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024, 5,674,713 and 5,700,673.

Specifications

Quantity 100 mL
Content And Storage 1 x 1 Vial Kinase-Glo™ substrate, 1 x 100mL Kinase-Glo™ Buffer -20°C
Detection Method Homogeneous, Non-radioactive Method
For Use With (Application) For determining the activity of purified Kinases by quantifying the amount of ATP
Product Type Kinase-Glo Luminescent Kinase Assay
For Use With (Equipment) For multiwell plates