Learn More
Promega ImProm-II™ Reverse Transcription DFS Item
Formulated for stand-alone applications or for easy transition to gene-specific target amplification.
Supplier:  Promega A3803
 View more versions of this product
Catalog No. PR-A3803
Only null left
Explore available promotions
Description
Description
  • Reverse transcribes RNA templates from total RNA, poly(A)+ mRNA, or synthetic transcript RNA
  • Enables robust, full-length cDNA synthesis for the analysis of rare or long messages (up to 8.9kb)
  • Uses volumes of 1–20μL from the initial RT reaction in coupled or uncoupled PCR reactions
  • May also be used for the incorporation of regular, Cy™3-modified, Cy5-modified, and amino-allyl-modified nucleotides for microarray applications
  • Refill components available separately
Specifications
Specifications
-30°C to -10°C
MgCl2 3 × 1.2mL, ImProm-II™ 5X reaction buffer 2 × 1250μL, ImProm-II reverse transcriptase 500μL
7
Liquid
Reverse transcription of RNA templates from total RNA, poly(A)+ mRNA, or synthetic transcript RNA
500
ImProm-II Reverse Transcriptase
Product Suggestions
Product Suggestions