missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega E. coli S30 T7 High-Yield Protein Expression System
The E. coli S30 T7 High-Yield Protein Expression System is designed to express up to 500ug/ml of protein in 1 hour from plasmid vectors containing a T7 promoter and a ribosome binding site.
$432.17 - $1088.33
Specifications
| Test Time | 1 hr. |
|---|---|
| Yield | 500 μg/mL |
| Type | E. coli Protein Expression System |
| Content And Storage | <-65°C |
| For Use With (Application) | Protein-Protein interactions, Protein-DNA interactions, Enzyme activity, Antibody production |
Includes: T7 S30 Extract, an optimized S30 Premix Plus provides all other required components, including amino acids, rNTPs, tRNAs, an ATP-regenerating system, IPTG and appropriate salts to express high levels of recombinant proteins. Also includes S30 T7 Control DNA, a control DNA template.
Description
- Simplified Transcription/Translation of DNA Cloned in Plasmid or Lambda Vectors with a T7 Promoter
- Express up to 500μg/mL of protein in one hour
- Flexible detection of expressed proteins by Coomassie(R) staining or fluorescence or biotinylated labels
Specifications
| 1 hr. | |
| E. coli Protein Expression System | |
| Protein-Protein interactions, Protein-DNA interactions, Enzyme activity, Antibody production |
| 500 μg/mL | |
| <-65°C |
U.S. Patents 5,292,658, and 5,418,155; Canadian Patent 2,105,984 and related patents.