Learn More
Promega GoTaq™ qPCR Master Mix DFS Item
GoTaq™ qPCR Master Mix is a ready-to-use, stabilized 2X formulation containing GoTaq(R) Hot Start Polymerase, MgCl2, dNTPs, BRYT Green(R) Dye and a proprietary reaction buffer.
Supplier:  Promega C5411
 View more versions of this product
Catalog No. PRC5411
Only null left
Explore available promotions
Includes: Tube of 100X CXR Reference Dye for use with instruments that require a higher level of reference dye than that in the GoTaq qPCR Master Mix
Description
Description
  • Efficient qPCR for Sensitive Amplification
  • Ready-to-use mix of optimized components for robust amplification
  • Compatible with both fast and standard cycling methods
  • Resistant to a wide range of PCR inhibitors
These catalog numbers are not available in Europe. Use of these products in all other countries except the U.S. is covered by one or more of the following U.S. patents and corresponding patent claims outside the U.S.: 5,079,352; 5,789,224; 5,618,711; 6,127,155 and claims outside the U.S. corresponding to U.S. Patent No. 4,889,818. The purchase of this product includes a limited, nontransferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser's own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser's activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404 USA. Use of these products in the U.S. for basic PCR is outside of any valid U.S. patents assigned to Hoffman La-Roche or Applera.
These products can be used in the U.S. for basic PCR in research, commercial or diagnostic applications without any license or royalty fees.
Specifications
Specifications
• CXR Reference Dye, 1 x 100 μL
Store at −30 to −10°C.
100 μL
Real Time PCR (qPCR)
Reference Dye
Product Suggestions
Product Suggestions