missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ Hot Start Polymerase: Green Master Mix, 2X DFS Item

GoTaq™ Hot Start Master Mixes are all-in-one master mixes (just add template and primers) that contain the high-performance GoTaq™ DNA Polymerase bound to a proprietary antibody that blocks polymerase activity until the reaction is heated at 94-95°C for two

Supplier:  Promega M5122

 View more versions of this product

Catalog No. PR-M5122


Only null left
Explore available promotions

Includes: GoTaq Hot Start Polymerase, buffer, dNTPs, and MgCl2, and supplied with nuclease-free water.

Description

Description

  • Hot-Start GoTaq(R) DNA Polymerase in a Ready-to-Use Master Mix
  • All-in-one master mix: just add template and primers
  • Minimize nonspecific amplification and primer-dimers
  • Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required

  • U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
  • Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.

Specifications

Specifications

5 U/μL
RT-PCR
100
2X Hot start green master mix (2 x 1250μL), Nuclease free Water (2 x 1250μL). Store at -20°C
2 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions