missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega GoTaq™ Hot Start Polymerase: Green Master Mix, 2X
Description
Includes
GoTaq Hot Start Polymerase, buffer, dNTPs, and MgCl2, and supplied with nuclease-free water.
- Hot-Start GoTaq(R) DNA Polymerase in a Ready-to-Use Master Mix
- All-in-one master mix: just add template and primers
- Minimize nonspecific amplification and primer-dimers
- Use the green master mix for direct-to-gel analysis, and the colorless master mix when post-amplification fluorescence or absorbance analysis is required
- U.S. Patent No. 6,242,235, Australian Patent No. 761757, Canadian Patent No. 2,335,153.
- Licensed under U.S. Patent Nos. 5,338,671 and 5,587,287 and corresponding patents in other countries.
Specifications
Specifications
| Concentration | 5 U/μL |
| Content And Storage | 2X Hot start green master mix (1 x 25mL), Nuclease free Water (1 x 25mL). Store at -20°C |
| For Use With (Application) | Cloning |
| Activation Time | 2 min. |
| No. of Reactions | 1000 |