missing translation for 'onlineSavingsMsg'
Learn More

Promega GoTaq™ DNA Polymerase

Taq DNA Polymerase. Provided with Green and Colorless Reaction Buffers. Green Buffer allows direct gel analysis after amplification. Enhanced performance over standard Taq DNA Polymerase.

Supplier:  Promega M3008

Encompass

 View more versions of this product

Catalog No. PRM3008


Only null left

Includes:
  • GoTaq DNA Polymerase; 5X Colorless GoTaq Reaction Buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction); 5X Green GoTaq Reaction Buffer (Contains two dyes that separate during electrophoresis to facilitate direct-to-gel analysis of PCR products). Both buffers are supplied at pH 8.5 and contain MgCl2 at a concentration of 7.5mM, giving a final concentration of 1.5mM in a 1X reaction
  • GoTaq Green Master Mix or GoTaq Colorless Master Mix, nuclease-free water

Description

Description

  • Taq DNA Polymerase with Buffers for Enhanced Amplification
  • Direct-to-gel convenience
  • Can be used with your current cycling conditions
  • Supplied at a concentration of 5u/μL
Specifications

Chemical Identifiers

5 U/μL
>90% Pure

Specifications

5 U/μL
>90% Pure
Standard PCR, RT-PCR
• GoTaq™ DNA Polymerase
• 5X Green GoTaq™ Reaction Buffer, 4 x 5 mL
• 5X Colorless GoTaq™ Reaction Buffer, 4 x 5 mL
• 7.5 mM MgCl2
• Nuclease-free Water

Store at –30° to –10°C.
2500 U
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions