Learn More
Promega GoTaq™ DNA Polymerase
Taq DNA Polymerase. Provided with Green and Colorless Reaction Buffers. Green Buffer allows direct gel analysis after amplification. Enhanced performance over standard Taq DNA Polymerase.
Supplier: Promega M3001
Includes:
- GoTaq DNA Polymerase; 5X Colorless GoTaq Reaction Buffer (Used when direct fluorescence or absorbance readings are required without prior purification of the DNA from the amplification reaction); 5X Green GoTaq Reaction Buffer (Contains two dyes that separate during electrophoresis to facilitate direct-to-gel analysis of PCR products). Both buffers are supplied at pH 8.5 and contain MgCl2 at a concentration of 7.5mM, giving a final concentration of 1.5mM in a 1X reaction
- GoTaq Green Master Mix or GoTaq Colorless Master Mix, nuclease-free water
Description
- Taq DNA Polymerase with Buffers for Enhanced Amplification
- Direct-to-gel convenience
- Can be used with your current cycling conditions
- Supplied at a concentration of 5u/μL
Chemical Identifiers
| 5 U/μL | |
| >90% Pure |
Specifications
| 5 U/μL | |
| >90% Pure | |
| RT-PCR |
| • GoTaq™ DNA Polymerase • 5X Green GoTaq™ Reaction Buffer, 1 mL • 5X Colorless GoTaq™ Reaction Buffer, 1 mL • 7.5 mM MgCl2 • Nuclease-free Water Store at –30° to –10°C. |
|
| 100 U |