Learn More
Promega Fetuin DFS Item
Fetuin is a glycoprotein with O-linked and N-linked glycosylation sites.
Supplier:  Promega V4961
Catalog No. FSTP9780272
Only null left
Explore available promotions
Description
Description
  • Glycoprotein with O- and N-Linked Glycosylation Sites
  • Deglycosylation substrate control for PNGase F, EndoH activity monitoring
  • Supplied at a concentration of 10mg/ml
Specifications
Specifications
10 mg/mL
1 x 50μL Fetuin
500 μg
Determining the state of Protein Glycosylation, characterizing Glycan structure
-30°C to -10°C
Fetuin
Product Suggestions
Product Suggestions