missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Genomic DNA
Purified genomic DNA. Greater than 90% of the DNA is longer than 50kb in size.
$372.88 - $445.35
Specifications
| Product Type | Genomic DNA |
|---|---|
| Storage Buffer | 10mM Tris HCl (pH 8), 1mM EDTA |
| Content And Storage | 2°C to 10°C |
| Quantity | 100 μg |
| For Use With (Application) | Southern blotting hybridizations, genomic analysis (including PCR) and genomic library construction |
| Catalog Number | Mfr. No. | Quantity | Source | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Source | Price | Quantity | ||||
PR-G1471
|
Promega
G1471 |
100 μg | Male Human |
Each for $372.88
|
|
||||
PR-G1521
|
Promega
G1521 |
100 μg | Female Human |
Each for $372.88
|
|
||||
PR-G3091
|
Promega
G3091 |
100 μg | Swiss-Webster albino mice |
Each for $445.35
|
|
||||
PR-G3041
|
Promega
G3041 |
100 μg | Human |
Each for $372.88
|
|
||||
Description
- Greater than 90% of the DNA is longer than 50kb in size, as measured by pulsed-field gel electrophoresis
- Mouse Genomic DNA is isolated from whole blood from disease-free mice
- Human Genomic DNA comes from multiple anonymous donors
- Storage Buffer: 10mM Tris-HCl (pH 8.0), 1mM EDTA
Specifications
| Genomic DNA | |
| 2°C to 10°C | |
| Southern blotting hybridizations, genomic analysis (including PCR) and genomic library construction |
| 10mM Tris HCl (pH 8), 1mM EDTA | |
| 100 μg |
PR-G1471, PR-G1521, PR-G3041 For Laboratory Use