missing translation for 'onlineSavingsMsg'
Learn More

Promega Genomic DNA DFS Item

Purified genomic DNA. Greater than 90% of the DNA is longer than 50kb in size.

Supplier:  Promega G3091

 View more versions of this product

Catalog No. PR-G3091


Only null left
Explore available promotions

Description

Description

  • Greater than 90% of the DNA is longer than 50kb in size, as measured by pulsed-field gel electrophoresis
  • Mouse Genomic DNA is isolated from whole blood from disease-free mice
  • Human Genomic DNA comes from multiple anonymous donors
  • Storage Buffer: 10mM Tris-HCl (pH 8.0), 1mM EDTA

Specifications

Specifications

Genomic DNA
10mM Tris HCl (pH 8), 1mM EDTA
100 μg
Swiss-Webster albino mice
2°C to 10°C
Southern blotting hybridizations, genomic analysis (including PCR) and genomic library construction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

PR-G1471, PR-G1521, PR-G3041 For Laboratory Use