missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Genomic DNA
Description
- Greater than 90% of the DNA is longer than 50kb in size, as measured by pulsed-field gel electrophoresis
- Mouse Genomic DNA is isolated from whole blood from disease-free mice
- Human Genomic DNA comes from multiple anonymous donors
- Storage Buffer: 10mM Tris-HCl (pH 8.0), 1mM EDTA
Specifications
Specifications
| Product Type | Genomic DNA |
| Source | Human |
| Storage Buffer | 10mM Tris HCl (pH 8), 1mM EDTA |
| Content And Storage | 2°C to 10°C |
| Quantity | 100 μg |
| For Use With (Application) | Southern blotting hybridizations, genomic analysis (including PCR) and genomic library construction |
PR-G1471, PR-G1521, PR-G3041 For Laboratory Use