Learn More
Promega Beetle Luciferin, Potassium Salts DFS Item
Beetle luciferin (also known as D-luciferin) is synthesized as the monopotassium salt and is a substrate for the beetle luciferase reporter systems.
Supplier:  Promega E1601
 View more versions of this product
Catalog No. PR-E1601
Only null left
Explore available promotions
Description
Description
  • Use in solution assays in which the production of light can be monitored with either a luminometer or a scintillation counter
  • Can penetrate cell membranes, allowing transformed cells to be monitored for luciferase activity
Specifications
Specifications
1 x 5mg Beetle Luciferin, Potassium salt
<-65°C
Fluid
318.4
5 mg
Product Suggestions
Product Suggestions