missing translation for 'onlineSavingsMsg'
Learn More

Promega Beetle Luciferin, Potassium Salts DFS Item

Beetle luciferin (also known as D-luciferin) is synthesized as the monopotassium salt and is a substrate for the beetle luciferase reporter systems.

$144.93 - $1285.97

Specifications

Molecular Weight (g/mol) 318.4
Content And Storage <-65°C
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-E1602 DFS Item
View Documents
Promega
E1602
50 mg
Each for $383.43
Only null left
 
PR-E1601 DFS Item
View Documents
Promega
E1601
5 mg
Each for $144.93
Only null left
 
PR-E1603 DFS Item
View Documents
Promega
E1603
250 mg
Each for $801.11
Only null left
 
PRE1605 DFS Item
View Documents
Promega
E1605
1 g
Each for $1,285.97
Only null left
 
Description

Description

  • Use in solution assays in which the production of light can be monitored with either a luminometer or a scintillation counter
  • Can penetrate cell membranes, allowing transformed cells to be monitored for luciferase activity
Specifications

Specifications

318.4
<-65°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'