missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ProK
Description
ProK is a lysine-based unnatural amino acid (UAA) used for genetic code expansion. It enables bioconjugation of fluorescent probes in E. coli or multicellular organisms.
Spécifications
Spécifications
| Chemical Name or Material | N6-[(2-Propyn-1-yloxy)carbonyl]-L-lysine hydrochloride |
| CAS | 1428330-91-9 |
| Quantity | 50 mg |
| Target | Unnatural Amino Acids |
| Molecular Formula | C10H16N2O4.HCl |
| Purity | 0.95 |