missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ ProCulture Warming Plate, Ambient to 37°C, Aluminum

Catalog No. 14513613
Change view
Click to view available options
Voltage:
120 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
14513613 120 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14513613 Supplier Bel-Art™ Supplier No. F370150000
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Warming plate is rectangular in shape and is highly accuracte outside the incubator.

  • Increase cell viability during cell passaging with this highly accurate warming plate that keeps cells near an optimal 37°C (±1°C) outside of the incubator.
  • Avoids numerous entries into the CO2 incubator which alter its atmosphere, harming ongoing experiments in the incubated cultures
  • Reduces cell stress and allows enzymes like trypsin to work efficiently
  • Can be used to maintain the warmth of reagents inside the hood
  • Fits easily in cell culture hoods or on the workbench
  • Temperature remains uniform across the entire anodized aluminum warming plate; measures 17.8 D x 20.3 cm W (7 x 8 in.)
  • 15-minute count down timer keeps track of time sensitive applications
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • Water resistant; safe for cleaning with Ethanol or IPA
  • 220V is CE marked

Compliance

UL approval pending.

Specifications

Top Plate Material Aluminum
Size 7 x 8 in.
Temperature (Metric) Ambient to 37°C
Depth (English) 10 in.
Depth (Metric) 25.4 cm
Height (English) 2.25 in.
Width (English) 9.75 in.
Height (Metric) 5.7 cm
Width (Metric) 24.9 cm
Wattage 22 w
Electrical Requirements 120 V, 50/60 Hz
Frequency 50/60 Hz
Plug Type US
Shape Rectangle
Voltage 120 V
Type Warming Plate
Show More Show Less