missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Probucol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 128°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 516.84 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Probucol |
Safety and Handling
| RTECSNumber | AL3705000 |
| TSCA | No |