missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Probenecid

Catalog No. 410750
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
410750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 410750 Supplier R&D Systems™ Supplier No. 4107/50

missing translation for 'validMainframeMsg'

A Tocris product, part of R&D Systems. MRP inhibitor

Probenecid is an inhibitor of multidrug resistance-associated proteins (MRP). Probenicid inhibits OAT3, organic acid transport in the kidney and other organs and exhibits inhibitory activity against pannexin 1 channels (IC50 ∼ 150 μM). Probenecid potently inhibits SARS-CoV-2 replication in NHBE and Vero E6 cells (IC50 = 1.3 nM and 750 nM, respectively) and reduces lung virus titers in vivo. Probenicid also inhibits influenza A virus replication in vitro and in vivo.

Chemical Identifiers

CAS 57-66-9
Molecular Formula C13H19NO4S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 285.36
Purity >98%
Molecular Weight (g/mol) 285.36
Chemical Name or Material Probenecid
Recommended Storage Store at Room Temperature