Learn More
Pro-Lab Diagnostics™ Mucolyse™ Sputum Digestant
Catalog No. 23265635
Encompass
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
23265635 100 mg
1 options
Catalog No. 23265635 Supplier Pro-Lab Diagnostics™ Supplier No. PL.701
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to liquefy sputum rapidly
  • Provides a simple, quantitative method for evaluating sputum bacteriology
  • Each vial can be reconstituted with distilled water to make 100mL of solution
Order Info
Packaged 10 vials per box
Specifications
Product Type Mucolyse Sputum Digetant
Includes 100 mg Dithiothreitol, Phosphate Buffer
Quantity 100 mg
Storage Requirements 2°C to 8°C