Learn More
United Scientific Supplies Prism and Lens Sets
Catalog No. S90324
Change view
Click to view available options
Material:
Acrylic
Catalog No. Material
S90324 Acrylic
1 options
Catalog No. S90324 Supplier United Scientific Supplies Supplier No. OPSETP3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Teach a wide variety of optical principles.
Set housed in a wooden case. Shipping weight: 2 lb. (0.9kg); box dimensions: 9.25 x 8 x 2 in. (23.1 x 20 x 5cm). Glass and acrylic sets each have seven components.
  • Double convex lens
  • Double concave lens (can be used together with double convex lens)
  • Semicircular lens
  • Trapezoidal prism
  • Small right-angle prism
  • Equilateral prism
  • Large right-angle refraction prism
Specifications
Material Acrylic
For Use With (Application) Teach a wide variety of optical principles
Product Type Prism and Lens Set
Includes Double convex lens, double concave lens, semicircular lens, trapezoidal prism, small right angle prism, equilateral prism, large right angle refraction prism