Learn More
Brady™ Magnetic Vinyl Sign: FULL CYLINDER
Catalog No. 19107762
Change view
Click to view available options
Dimensions (L x W):
5 x 7 in.
Catalog No. Dimensions (L x W)
19107762 5 x 7 in.
1 options
Catalog No. 19107762 Supplier Brady™ Supplier No. SM571E
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Improve workplace safety with proper signage and markings. This magnetic Brady™ vinyl sign is a durable, clear and effective tool to raise awareness to workplace hazards.
  • Magnetic vinyl material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Self sticking vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics
  • Excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Material Vinyl
Mounting Type Magnetic
Shape Rectangular
Languages English
Dimensions (L x W) 5 x 7 in.
Displayed Text FULL CYLINDER
Product Type Safety Sign
Color Green on White