missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Cable Lockout Device

Catalog No. 19103938
Change view
Click to view available options
Includes:
10 ft. cable
6 ft. cable
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19103938 10 ft. cable
19103937 6 ft. cable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19103938 Supplier Brady™ Supplier No. CABLO10

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lockout gates, handles, valves and more! The Brady™ Cable Lockout Device is a versatile solution for all of your facility's lockout needs.

  • Versatile device for all of your unusual energy isolating devices that are difficult to lock out
  • 6 lock holes in the handle are 9/32 in. in diameter, which accommodates most lock shackles and hasps
  • Includes 10 in. cable

Specifications

Product Type Tab-cinching Cable Lockout Device
Color Red
Material Plastic
Dimensions (L x W x H) 0.65 x 5.84 x 2 in.
Includes 10 ft. cable
Quantity 1 Ea.