Learn More
Beckman Coulter 460514, Rotor, A-4-38, Swing Bucket, 6 x 85ml
Numéro de catalogue. NC0696442
Encompass
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 xg. The rotor capacity is 18 x 1.5/2.0mL tubes. It has a compact footprint, digital display, 10 acceleration and braking rates, and a pulse key for short spin cycles. The Microfuge 18 is ideal for microcentrifuge applications in clinical, research, and testing labs.
Numéro de catalogue. NC0696442 Fournisseur Beckman Coulter Code fournisseur BEC460514
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 xg. The rotor capacity is 18 x 1.5/2.0mL tubes. It has a compact footprint, digital display, 10 acceleration and braking rates, and a pulse key for short spin cycles. The Microfuge 18 is ideal for microcentrifuge applications in clinical, research, and testing labs.