missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genlantis™ PrimaPure™ Fibroblasts
High-quality human adult, fetal, and neonatal fibroblasts
$1,379.75 - $2,982.86
Specifications
| For Use With (Application) | For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing |
|---|---|
| Media Recommendation | Fibroblast Growth Medium |
| Quantity | >500000 Cells |
| Sufficient For | 16 Population Doublings |
| Catalog Number | Mfr. No. | Cell Type | Source | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Cell Type | Source | Price | Quantity & Availability | ||||
PH10605A
|
Genlantis™
PH10605A |
Adult HDF | Human Adult Skin |
Each for $2,982.86
|
|
||||
PH10605F
|
Genlantis™
PH10605F |
Fetal HDF | Human Fetal |
Each for $1,739.19
|
|
||||
PH10605N
|
Genlantis™
PH10605N |
Neonatal HDF | Human Neonatal Foreskin |
Each for $1,379.75
|
|
||||
Description
Each lot functionally characterized
Single donors to allow variability comparisons
Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi
16 population doublingsguaranteed with Genlantis media and reagents
Fibroblasts are cells that make the structural fibers and ground substance of connective tissue.
Specifications
| For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing | |
| Fibroblast Growth Medium | |
| >500000 Cells | |
| 16 Population Doublings |