Learn More
Genlantis™ PrimaPure™ Fibroblasts
High-quality human adult, fetal, and neonatal fibroblasts
Supplier: Genlantis™ PH10605F
Description
Each lot functionally characterized
Single donors to allow variability comparisons
Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi
16 population doublingsguaranteed with Genlantis media and reagents
Fibroblasts are cells that make the structural fibers and ground substance of connective tissue.
Chemical Identifiers
| For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing | |
| Fibroblast Growth Medium | |
| >500000 Cells | |
| 16 Population Doublings |
Specifications
| Fetal HDF | |
| Fibroblast Growth Medium | |
| Human Fetal |
| For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing | |
| >500000 Cells | |
| 16 Population Doublings |