missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Fibroblasts

Catalog No. PH10605A
Encompass
Change view
Click to view available options
Cell Type:
Adult HDF
Fetal HDF
Neonatal HDF
Source:
Human Adult Skin
Human Fetal
Human Neonatal Foreskin
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type Source
PH10605A Adult HDF Human Adult Skin
PH10605F Fetal HDF Human Fetal
PH10605N Neonatal HDF Human Neonatal Foreskin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PH10605A Supplier Genlantis™ Supplier No. PH10605A

High-quality human adult, fetal, and neonatal fibroblasts

Each lot functionally characterized

Single donors to allow variability comparisons

Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi

16 population doublingsguaranteed with Genlantis media and reagents
Fibroblasts are cells that make the structural fibers and ground substance of connective tissue.

Specifications

Cell Type Adult HDF
For Use With (Application) For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing
Media Recommendation Fibroblast Growth Medium
Quantity >500000 Cells
Source Human Adult Skin
Sufficient For 16 Population Doublings