missing translation for 'onlineSavingsMsg'
Learn More

Genlantis™ PrimaPure™ Fibroblasts

Catalog No. PH10605N
Encompass
Change view
Click to view available options
Cell Type:
Fetal HDF
Neonatal HDF
Source:
Human Fetal
Human Neonatal Foreskin
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type Source
PH10605N Neonatal HDF Human Neonatal Foreskin
PH10605F Fetal HDF Human Fetal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PH10605N Supplier Genlantis™ Supplier No. PH10605N

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-quality human adult, fetal, and neonatal fibroblasts

Each lot functionally characterized

Single donors to allow variability comparisons

Each lot tested negative for HIV, Hepatitis B C, mycoplasma, bacteria, yeast, and fungi

16 population doublingsguaranteed with Genlantis media and reagents
Fibroblasts are cells that make the structural fibers and ground substance of connective tissue.

Specifications

Cell Type Neonatal HDF
For Use With (Application) For in-vitro analysis of fibroblast growth, migration and collagen metabolism in wound healing
Media Recommendation Fibroblast Growth Medium
Quantity >500000 Cells
Source Human Neonatal Foreskin
Sufficient For 16 Population Doublings