missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prilocaine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 166°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 256.77 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Prilocaine Hydrochloride |
Safety and Handling
| RTECSNumber | UG5775000 |
| TSCA | No |