missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences Prep Desalting Column
Description
- Prep Desalting columns are mostly used for removal of salt or buffer exchange before or after different chromatographic steps.
- Prep Desalting columns can be used for separation of proteins based on their sizes.
- G‐Biosciences offer different sizes of desalting columns.
- Prep Desalting columns are made of biocompatible polypropylene, which does not interact with biomolecules.
- The column has a stopper at the inlet and one top plug, one end cap, two filter discs.
Spécifications
Spécifications
| Product Type | Desalting Column |
| Format | Gravity Flow Column |
| Column Type | Size-exclusion, Proprietary Resin |
| MWCO | 5.0 kDa |
| Product Line | Pierce |
| Disposable | Reusable |
| Quantity | 5 x 150 mL |
| Sample Volume (Metric) | 150 mL |