Learn More
Eisco™ Premium Square Head Boss Head Clamp
Catalog No. S13923
Change view
Click to view available options
Color:
Blue
Catalog No. Color
S13923 Blue
1 options
Catalog No. S13923 Supplier Eisco™ Supplier No. PRBH05
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Premium Square Head Boss Head Clamp able to hold rods up to 15 mm in diameter.
  • Threaded metal one piece clamping screws
  • Able to hold rods up to 15 mm in diameter
  • Holds rods orthogonal (90 degrees) to one another
  • All metal constructions to carry heavy loads and prevent torque damage
Specifications
Autoclavable Autoclavable
Type Bosshead Clamp
Material Nickel-plated
Holds Rods up to 15 mm diameter
Shape Square
Color Blue
Jaw Type 1-Prong