missing translation for 'onlineSavingsMsg'
Learn More

Mopec Prefilled Formalin Cube - 5 Gallons - 20 NBF

Catalog No. 21010271
Change view
Click to view available options
Form:
Liquid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21010271 Liquid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21010271 Supplier Mopec Supplier No. BG52520
Only null left

Mopec's Bulk Formalin Containers are filled with a 10% neutral buffered formalin used as a general purpose fixative for tissues. Penetrates quickly, but fixes slowly. No other dilution or addition of other agents is necessary. Comes in a variety of sizes.

  • Filled with a 10% neutral buffered formalin
  • General purpose fixative for tissues
  • Penetrates quickly, but fixes slowly
  • No other dilution or additional agents are necessary
  • Sold individually

Specifications

For Use With (Equipment) All Tissue Processors
Grade Professional Grade
Product Type Collection Fluid
Solution Type Formalin
Form Liquid