missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Precoated TLC and PLC Glass Plates

Catalog No. M1057210001
Change view
Click to view available options
Sorbent:
Cellulose
Cellulose F
Kieselguhr F254
PEI Cellulose F
Silica Gel 60
Silica Gel 60 F254
Silica Gel 60 F254 AMD, extra thin
Silica Gel 60 F254 with preconcentrating zone
Silica Gel 60 RP-18 F254
Silica Gel 60 RP-18 F254 s
Silica Gel 60 RP-2 (silanized)
Silica Gel 60 RP-2 F254 (silanized)
Silica Gel 60 RP-8 F254
Silica Gel 60 RP-8 F254 s
Silica Gel 60 W F254 s
Silica Gel 60 with preconcentrating zone
Particle Size:
10 to 12 μm
5 to 20 μm
5 to 6 μm
15 μm
Dimensions (L x W):
2.5 x 7.5 cm
5 x 10 cm
5 x 20 cm
10 x 20 cm
20 x 10 cm
20 x 20 cm
Quantity:
100/Pk.
12/Pk.
20/Pk.
25/Pk.
50/Pk.
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sorbent Particle Size Dimensions (L x W) Quantity
M1057210001 Silica Gel 60 10 to 12 μm 20 x 20 cm 25/Pk.
M1153890001 Silica Gel 60 RP-18 F254 s 10 to 12 μm 20 x 20 cm 25/Pk.
M1117980001 Silica Gel 60 F254 with preconcentrating zone 5 to 20 μm 20 x 20 cm 25/Pk.
M1153270001 Silica Gel 60 F254 10 to 12 μm 2.5 x 7.5 cm 100/Pk.
M1156830001 Silica Gel 60 RP-18 F254 s 5 to 20 μm 5 x 20 cm 50/Pk.
M1118460001 Silica Gel 60 F254 with preconcentrating zone 10 to 12 μm 10 x 20 cm 50/Pk.
M1057450001 Silica Gel 60 5 to 20 μm 20 x 20 cm 12/Pk.
M1117640001 Silica Gel 60 F254 AMD, extra thin 5 to 6 μm 20 x 10 cm 25/Pk.
M1164850001 Silica Gel 60 W F254 s 10 to 12 μm 20 x 20 cm 25/Pk.
M1057250001 PEI Cellulose F 5 to 20 μm 20 x 20 cm 25/Pk.
M1057180001 Cellulose F 15 μm 20 x 20 cm 25/Pk.
M1154240001 Silica Gel 60 RP-8 F254 10 to 12 μm 10 x 20 cm 50/Pk.
M1057460001 Silica Gel 60 RP-2 (silanized) 10 to 12 μm 20 x 20 cm 25/Pk.
M1153260001 Silica Gel 60 10 to 12 μm 2.5 x 7.5 cm 100/Pk.
M1057380001 Kieselguhr F254 10 to 12 μm 20 x 20 cm 25/Pk.
M1154230001 Silica Gel 60 RP-18 F254 s 10 to 12 μm 10 x 20 cm 50/Pk.
M1156840001 Silica Gel 60 RP-8 F254 s 10 to 12 μm 5 x 10 cm 25/Pk.
M1138940001 Silica Gel 60 15 μm 20 x 20 cm 20/Pk.
M1057280001 Cellulose F 15 μm 10 x 20 cm 50/Pk.
M1057240001 Silica Gel 60 5 to 20 μm 5 x 20 cm 100/Pk.
M1118450001 Silica Gel 60 with preconcentrating zone 10 to 12 μm 20 x 20 cm 25/Pk.
M1057470001 Silica Gel 60 RP-2 F254 (silanized) 10 to 12 μm 20 x 20 cm 25/Pk.
M1153880001 Silica Gel 60 RP-8 F254 s 10 to 12 μm 20 x 20 cm 25/Pk.
M1057160001 Cellulose 15 μm 20 x 20 cm 25/Pk.
M1118440001 Silica Gel 60 with preconcentrating zone 15 μm 10 x 20 cm 50/Pk.
M1056260001 Silica Gel 60 10 to 12 μm 10 x 20 cm 50/Pk.
M1057300001 Cellulose 15 μm 10 x 20 cm 50/Pk.
M1156850001 Silica Gel 60 RP-18 F254 10 to 12 μm 20 x 20 cm 25/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
Catalog No. M1057210001 Supplier MilliporeSigma™ Supplier No. 1.05721.0001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Average particle size is 15μm with a 5-20μm distribution

  • Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
  • 60 Å silica gel is suitable for all common gel separations including weak to strong polar analytes
  • Silica can also be used for applications requiring charring procedures
  • Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
  • For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates

Certifications

ISO 9001:2000

†With acid-stable fluorescent indicator.

‡PEI (polyethyleneimine) should be stored at 4°C (39°F). If stored at room temperature, PEI plates may turn slightly yellow, which could affect separation. To minimize yellow color, predevelop in distilled water.

Specifications

Backing Material Glass
Material Silica 60
Particle Size 10 to 12 μm
Pore Size 60 Å
Quantity 25/Pk.
Type Unmodified Silica Gel TLC Plate
Fluorescent Indicator No
For Use With (Application) In-Process Control in Drug Synthesis, Identity and Stability Testing of Drugs, Quality Control of Pharmaceuticals, Food and Environmental Samples and Residue Analysis in Food and Environmental Samples
Sorbent Silica Gel 60
Thickness (Metric) 250 μm
Dimensions (L x W) 20 x 20 cm
Length (Metric) 20 cm
Width (Metric) 20 cm
Show More Show Less