missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Precision™ Shaking Water Baths

Get shaking water baths that support a range of sensitive life science and QA/QC applications, from warming fragile reagents to tissue culture and genetic sequencing.

Supplier:  Thermo Scientific™ TSDUB15

 Afficher plus de versions de ce produit

Numéro de catalogue. TSDUB15

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.


Description

Description

Benefit from Thermo Scientific™ Precision™ shaking water baths, which support a range of sensitive life science and QA/QC applications – from warming fragile reagents to tissue culturing and genetics sequencing. Easily clean and maintain your bath with new coil-free interior. The Precision shallow form shaking bath has a removable tray that allows for use with smaller sample containers. Precision Dubnoff shaking water baths are designed specifically for applications that require your samples to be incubated in a controlled atmosphere

  • Easily clean and maintain bath with coil-free design
  • Optimize scheduling with auto-on and auto-off timers
  • Help prevent bath overheating and damage with low-fluid protection
  • Conveniently save commonly used settings with four temperature and shaking speed presets
  • Help protect your work with audible alarms
  • Easily operate and monitor with icon-based graphical display
  • Accommodate taller labware with new hinged lid and extended height
  • Features next-generation shaker motor
Spécifications

Identifiants chimiques

304 Stainless Steel
UL, CE
Shaking Water Bath
Epoxy Powder-coated Cold Rolled Steel
±0.1°C
Global Voltage 100/115 V 200/230 V 50/60 Hz
Digital
Ambient +5° to 100°C
Monochrome LCD
4
±0.05°C

Spécifications

304 Stainless Steel
15 L
Ambient +5° to 100°C
Monochrome LCD
Large and Small Gassing Hood and Rubber Duck
Precision DUB 15
±0.1°C
25 x 15.6 x 9.9 in. (636 x 396 x 250 mm)
9.0-10.5 / 4.3-5.0 (120V/230V)
Digital
UL, CE
Shaking Water Bath
1200 w
Epoxy Powder-coated Cold Rolled Steel
4
±0.05°C
Global Voltage 100/115 V 200/230 V 50/60 Hz
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions