missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Precision Sampling Syringe Needles

Numéro de catalogue. p-7213668
Change view
Click to view available options
Needle Point Style:
Cone with Side Port
For Use With (Equipment):
VICI Series A
VICI Series A-2, C, D
Volume (Metric):
100 to 10,000 μL
25 to 10,000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Needle Point Style For Use With (Equipment) Volume (Metric)
111011559 Cone with Side Port VICI Series A 100 to 10,000 μL
111020934 Cone with Side Port VICI Series A-2, C, D 25 to 10,000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 111011559 Fournisseur MilliporeSigma™ Supelco™ Code fournisseur 22288
Il en reste null

Ideal for collection, trace analysis, and sample concentration of environmental contaminants

Spécifications

Material Stainless Steel (Tip)
Description Series A
Diameter (English) Inner Needle 0.012 in.
Diameter (English) Outer Needle 0.029 in.
For Use With (Application) Precision Sampling
Length (English) Needle 2.25 in.
Product Type Syringe Needle
Needle Point Number 5
Quantity 3 Pk.
Packaging 3 Pk.
Volume (Metric) 100 to 10,000 μL
Needle Point Style Cone with Side Port
For Use With (Equipment) VICI Series A
Afficher plus de résultats Afficher moins de résultats