Learn More
Excelta™ Precision Fine Tip Wire Cutters
Features set-screw to prolong cutting life and prevent over-travel of cutting blades
$485.56 - $558.22
Specifications
Grade Ultra Precision
Material 1% Carbon Tool Steel
Cut Type Optimum Flush
Grip Type ESD-safe soft foam cushion
Includes Set-screw, induction hardened cutting blades to RHC 63-65, dual leaf springs
View More Specs
Products 5
Catalog Number Mfr. No. Capacity (English) Length (English) Handle Type Head Style Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Length (English) Handle Type Head Style Price Quantity & Availability  
17467458
View Documents Promotion Details
Excelta™
7249E
0.001 to 0.02 in. 5.5 in. Standard Relieved head, fine tip
Each for $485.56
Only null left
17467430
View Documents Promotion Details
Excelta™
7149E
0.001 to 0.02 in. 5.75 in. Ergonomic long Small relieved, fine tip
Each for $527.06
Only null left
17467461
View Documents Promotion Details
Excelta™
7271E
0.001 to 0.01 in. 5.25 in. Standard Small
Each for $558.22
Only null left
17467432
View Documents Promotion Details
Excelta™
7170E
0.001 to 0.02 in. 6.25 in. Ergonomic long Medium
Each for $523.56
Only null left
17467433
View Documents Promotion Details
Excelta™
7170EA
0.001 to 0.02 in. 6.25 in. Ergonomic long Medium fine tip, Slim head
Each for $532.34
Only null left
Description
Description
  • Optimum flush cut
  • For cutting soft wire in tight places
  • Set-screw Stop prevents premature wear
  • Induction hardened carbon tool steel
  • Dual leaf springs
  • ESD safe grips
Specifications
Specifications
Ultra Precision
Optimum Flush
Set-screw, induction hardened cutting blades to RHC 63-65, dual leaf springs
1% Carbon Tool Steel
ESD-safe soft foam cushion